Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam APC/Cy7 Conjugation Kit, Lightning-Link, 3 x 100 μg

Catalog No. 502585614
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-5614 3 x 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-5614 Supplier Abcam Supplier No. AB1028593X100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labeling in less than 4 hours with 30 seconds hands-on time and 100% antibody recovery.

  • Labeling Time: Less than 4 hours
  • Hands-on Time: 30 seconds
  • Recovery: 100% antibody recovery

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Protein Labeling Reagent
Content And Storage Store at -20°C.
Label or Dye APC/Cy7
Shipping Condition Ambient
Target Molecule Antibody
Alternate Part Number ab102859
Format Kit
Detection Method Fluorescence
Quantity 3 x 100 μg
For Use With (Application) Flow Cytometry, Fluorescence Microscopy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.