Learn More
Cytek Biosciences Inc APC-CY7 MS CD4 (RM4-5) 25UG
Supplier:  Cytek Biosciences Inc 250042U025
Encompass_Preferred
APC-Cyanine7 Anti-Mouse CD4 (RM4-5) Isotype: Rat IgG2a, kappa Apps: FC Size: 25 ug
Catalog No. 50-105-4253
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.