Learn More
Cytek Biosciences Inc Antibodies APC-Cyanine7 Anti-Mouse CD5 (53-7.3) ..... Cat. No. 25-0051-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 250051U025
Encompass_Preferred
Antibodies APC-Cyanine7 Anti-Mouse CD5 (53-7.3) ..... Cat. No. 25-0051-U025 ..... Size 25 µgThe 53-7.3 monoclonal antibody is specific for mouse CD5, a 67 kDa protein that is expressed on most thymocytes, mature T cells and the B-1 subset of B cells. CD5 is a member of the scavenger receptor cysteine-rich protein superfamily and is a ligand for the C-type lectin CD72. CD5 is also known as Leu-1 and functions to modulate antigen receptor signaling on T and B cells.
Catalog No. 50-210-3108
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.