Learn More
Tonbo Bioscience Antibodies APC-Cyanine7 Anti-Mouse CD69 (H1.2F3) ..... Cat. No. 25-0691-U025 ..... Size 25 µg
Supplier:  Tonbo Bioscience 250691U025
Encompass_Preferred
Antibodies APC-Cyanine7 Anti-Mouse CD69 (H1.2F3) ..... Cat. No. 25-0691-U025 ..... Size 25 µgThe H1.2F3 antibody reacts with mouse CD69, a type II transmembrane glycoprotein also known as the Very Early Activation Antigen, EA-1, Leu23, Activation Inducer Molecule (AIM) and CLEC2C. CD69 is expressed as a 60 kDa disulfide-linked homodimer on activated T and B cells, NK cells, neutrophils and monocytes. Induction occurs rapidly upon activation. It is also constitutively expressed on platelets and a subset of thymocytes. CD69 acts as a co-stimulatory molecule involved in activation and proliferation of T cells, and may be a marker for thymocytes undergoing TCR-mediated positive selection.Co-stimulation with the H1.2F3 clone has been reported to enhance T cell and macrophage activation. Please choose the appropriate format for each application.
Catalog No. 50-210-3129
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.