Learn More
Biolegend APC/Fire™ 750 anti-human CD88 (C5aR)
Supplier:  Biolegend 344316
Encompass_Preferred
APC/Fire™ 750 anti-human CD88 (C5aR) [S5/1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 100 tests
Catalog No. 50-207-6648
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.