Learn More
Biolegend APC/Fire™ 750 anti-human IL-22
Supplier:  Biolegend 366713
Encompass_Preferred
APC/Fire™ 750 anti-human IL-22 [2G12A41]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-207-8281
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.