Learn More
Biolegend APC/Fire™ 750 anti-mouse IgG2a
Supplier:  Biolegend 407124
Encompass_Preferred
APC/Fire™ 750 anti-mouse IgG2a [RMG2a-62]; Isotype: Rat IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-9808
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.