Learn More
ABclonal Technology APC Rabbit anti-Human CD28 mAb
SDP
Supplier:  ABclonal Technology A26815200T
Encompass_Preferred
The protein encoded by this gene is essential for T-cell proliferation and survival cytokine production and T-helper type-2 development Several alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-08460
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.