Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC Rat Anti-Human Folate Receptors α/β (FRαβ), BD Pharmingen™

Catalog No. p-200020627 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569811 100 Tests
BDB569885 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569811 Supplier BD Biosciences Supplier No. 569811
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Folate Receptors α/β (FRαβ) Monoclonal antibody specifically detects Folate Receptors α/β (FRαβ) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Folate Receptors α/β (FRαβ)
Applications Flow Cytometry
Classification Monoclonal
Clone 5 (also known as No.5; No. 5; Antibody No.5; Antibody No. 5; Clone 5; 5/FRAB)
Conjugate APC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias FOLR1, Folate receptor 1, FR-alpha, FRalpha; FOLR2, Folate receptor 2, FR-beta, FRbeta; FRαβ, FR-αβ
Host Species Rat
Immunogen Human Folate Receptor beta Transfected Cell Line
Purification Method Affinity Chromatography
Quantity 100 Tests
Regulatory Status RUO
Gene ID (Entrez) 2348, 2350
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.