Learn More
AdipoGen Apcin (25 mg)
SDP
Supplier:  AdipoGen AGCR13603M025
Encompass_Preferred
Chemical. CAS: 300815-04-7. Formula: C13H14Cl3N7O4. MW: 438.7. Cell permeable compound capable of slowing anaphase-promoting complex/cyclosome (APC/C)-dependent proteolysis. Inhibitor of the ubiquitin ligase activity of the APC/C by inhibiting substrate binding to Cdc20. Distinct mechanism of action than proTAME. Blocks mitotic exit only modestly when used alone in cultured cells. A strong synergistic effect is observed when Apcin and proTAME are used simultaneously, providing a more robust mitotic arrest. This may be very beneficial when treating cells that do not efficiently convert proTAME to its active parent compound (TAME) or if experiments demand strong APC/C inhibition.
Catalog No. 50-205-3972
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.