Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ APE1 Redox Inhibitor
SDP

Catalog No. NBP149581
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149581 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149581 Supplier Novus Biologicals™ Supplier No. NBP149581

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

For use in research applications

Specifications

For Use With (Application) Inhibits redox activity of APE1
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Quantity 5 mg
Product Type APE1 Redox Inhibitor E3330
Inhibitors E3330

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.