Learn More
CPC Scientific Glp-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific APEL002B
Encompass
Pyroglutamyl form of apelin-13 that displays a high activity with an EC50 value of 0.30 nM.SEQUENCE: Glp-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Glp-RPRLSHKGPMPFMOLECULAR FORMULA: C69H108N21O16S1MOLECULAR WEIGHT: 1533.81STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [217082-60-5]SYNONYMS: (Pyr)-Apelin-13 (human, bovine, mouse, rat)RESEARCH AREA: AntimicrobialREFERENCES:K. Tatemoto et al., Biochem Biophy. Res. Commun., 251, 471 (1998)SKU(s): APEL-002A, APEL-002B
Catalog No. 50-210-8781
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.