Learn More
Bachem Apelin-13 (human, bovine, mouse, rat)
SDP
Supplier:  Bachem 4029109.0005
Encompass_Preferred
Apelin-13 (human, bovine, mouse, rat), H-Gln-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe-OH, 217082-58-1, C69H111N23O16S, Mr 1550.85, 5mg. Apelin-13, generated from apelin-36, is an endogenous peptide ligand for the human APJ receptor, a putative receptor protein related to the angiotensin receptor (AT1). Apelin-13 exerted an acidification-rate-promoting activity in the CHO cells expressing the APJ receptor with a median effective concentration (EC50) value of 0.37 nM. In a study of Valle et al. chronic central administration of apelin-13 over 10 days increased food intake, body weight, locomotor activity and body temperature in C57BL/6 mice.
Catalog No. 50-194-6755
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.