Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APEX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675646 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-646 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-646 Supplier Invitrogen™ Supplier No. PA141306
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Suggested positive control: Hela whole cell extract. Predicted cross-reactivity based on sequence identity (not validated): Mouse, Rat. Immunogen sequence: EELRALDSEG RALLTQHKI & GQAKTSEAKD EKELRTSF.

Apurinic/apyrimidinic (AP) sites occur frequently in DNA molecules by spontaneous hydrolysis, by DNA damaging agents or by DNA glycosylases that remove specific abnormal bases. AP sites are pre-mutagenic lesions that can prevent normal DNA replication so the cell contains systems to identify and repair such sites. Class II AP endonucleases cleave the phosphodiester backbone 5' to the AP site. This gene encodes a protein shown to have a weak class II AP endonuclease activity. Most of the encoded protein is located in the nucleus but some is also present in mitochondria. This protein may play an important role in both nuclear and mitochondrial base excision repair (BER).
TRUSTED_SUSTAINABILITY

Specifications

Antigen APEX2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.02% sodium azide
Gene APEX2
Gene Accession No. Q9UBZ4
Gene Alias AP endonuclease 2; AP endonuclease XTH2; Ape2; APEX nuclease (apurinic/apyrimidinic endonuclease) 2; APEX nuclease 2; APEX nuclease-like 2; APEX2; APEXL2; apurinic/apyrimidinic endodeoxyribonuclease 2; apurinic/apyrimidinic endonuclease 2; apurinic/apyrimidinic endonuclease-like 2; Apurinic-apyrimidinic endonuclease 2; C430040P13Rik; DNA-(apurinic or apyrimidinic site) lyase 2; RP5-884M20.3; XTH2; ZGRF2; zinc finger, GRF-type containing 2
Gene Symbols APEX2
Host Species Rabbit
Immunogen Peptides of human APEX2 (aa 122-140 and 438-455).
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27301
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.