Learn More
MyBioSource Polyclonal Human APEX2 Antibody
SDP
Supplier:  MyBioSource MBS92078310.08 ML
Encompass_Preferred
The APEX2 Polyclonal Antibody from MyBioSource is a rabbit polyclonal antibody raised against a partial recombinant APEX2 protein. APEX2 is a multifunctional DNA repair enzyme involved in base excision repair. This antibody detects APEX2 protein at around 35 kDa by Western blot. The immunogen is a recombinant protein of human APEX2. This antibody is tested and validated for WB, ELISA, and IHC applications. The recommended dilution for WB is 1:500-1:1000, for ELISA it is 1:500-1:1000, and for IHC it is 1:50-1:200. This antibody is 100% guaranteed with free lifetime replacement.
Catalog No. NC2287243
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.