Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA12010 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1-2010 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1-2010 Supplier Invitrogen™ Supplier No. PA12010
  • $625.73
    $481.65 / Each of 1

    Save $144.08

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-2010 detects APH-1 in human and mouse samples. PA1-2010 has been successfully used in immunofluorescent, immunoprecipitation, and Western blot procedures. By Western blot, this antibody detects a ~28 kDa protein representing APH-1 in mouse brain and kidney extracts. The PA1-2010 immunogen is a Synthetic peptide, KLH conjugated, corresponding to residues C (245) R R Q E D S R V M V Y S A L R I P P E D (265) of the C-Terminal domain of the long isoform of APH-1a. This polyclonal antibody has been referred to as, O2C2 in the reference listed above.

APH1 is an essential subunit of the gamma-secretase complex, an endoprotease complex that catalyzes the intramembrane cleavage of integral proteins such as Notch receptors and APP (beta-amyloid precursor protein). The gamma-secretase complex is minimally composed of a presenilin homodimer (PSEN1 or PSEN2), nicastrin (NCSTN), APH1 (APH1A or APH1B) and PEN2, although other components may exist. APH1 probably represents a stabilizing cofactor for the presenilin homodimer that promotes the formation of a stable complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APH1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.005% sodium azide
Gene APH1A
Gene Accession No. Q8BVF7, Q96BI3
Gene Alias 6530402N02Rik; anterior pharynx defective 1 homolog A; anterior pharynx defective 1a homolog; anterior pharynx defective 1a homolog (C. elegans); APH1; APH-1; aph-1 homolog A, gamma secretase subunit; aph-1 homolog A, gamma-secretase subunit; Aph1a; APH-1a; APH1A gamma secretase subunit; Aph-1alpha; CGI-78; Gamma-secretase subunit APH-1A; hypothetical protein LOC539247; presenilin-stabilization factor; PSF; RGD1311546; RP4-790G17.3; UNQ579/PRO1141
Gene Symbols APH1A
Host Species Rabbit
Immunogen Synthetic peptide, KLH conjugated, corresponding to residues C (245) R R Q E D S R V M V Y S A L R I P P E D (265) of human APH-1aL/CT.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226548, 51107
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.