Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ apHera™ C4 HPLC Guard Column, 5 μm Particle Size

Catalog No. 111008044 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric) Inner:
7.5 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
11-100-8044 7.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-8044 Supplier MilliporeSigma™ Supelco™ Supplier No. 56332AST

missing translation for 'validMainframeMsg'

apHera™ reversed columns were devoloped specifically to provide the superior advantages of both silica and polystyrene columns, without the disadvantages of either.

apHera™ reversed columns were devoloped specifically to provide the superior advantages of both silica and polystyrene columns, without the disadvantages of either. This was accomplished using a vinyl alcohol copolymer base that keeps the surface wetted even with high carbon loads. The porous structure has an average pore diameter large enough to produce ideal results for small analytes, peptides and small proteins. These columns equal silica based columns in separation efficiency with organic solvents but provide efficiency with buffered alkaline solutions not possible on silica. One of the most significant features is logicial elution order of alkylated bases where retention increases proportionately with increasing chain length.Features of apHera™ Reversed Phase Columns. Stable vinyl copolymer base. Stable pH range 2-12. 300 angstrom pore size. Stable in all organic solvents. Expanded applications: peptides, proteins. Efficient separation of any basic substance. Amenability to washing and alkaline solutions
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1/Pk.
Particle Size 5 μm
Length (Metric) 5 cm
Pore Size 300 Å
Type Guard Column
Diameter (Metric) Inner 7.5 mm
Phase Reversed Phase
For Use With (Application) HPLC
Product Line apHera

apHera is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.