Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Aphidicolin

Catalog No. p-7070067
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60236MCR 1 mg
AAJ60236LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60236MCR Supplier Thermo Scientific Chemicals Supplier No. J60236MCR
Only null left
Add to Cart

A DNA polymerase inhibitor, blocks the cell cycle at the early S-phase | CAS: 38966-21-1 | C20H34O4, C20H34O4 | 338.49 g/mol

A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38966-21-1
Molecular Formula C20H34O4
Synonym APC

Specifications

Quantity 1 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
Formula Weight 338.48
Melting Point 224°C
Solubility Soluble in DMSO or methanol. Insoluble in water. Store solutions at -20°
Product Type Aphidicolin
Form Cyrstalline powder
Beilstein 4689958
InChI Key NOFOAYPPHIUXJR-APNQCZIXSA-N
SMILES C[C@@]1(CO)[C@H](O)CC[C@@]2(C)[C@H]1CC[C@H]1C[C@@H]3C[C@]21CC[C@]3(O)CO
IUPAC Name (1S,2S,5R,6R,7R,10S,12R,13R)-6,13-bis(hydroxymethyl)-2,6-dimethyltetracyclo[10.3.1.0¹,¹⁰.0²,⁷]hexadecane-5,13-diol
Molecular Weight (g/mol) 338.49
Show More Show Less
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.