Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Aphidicolin, DNA Polymerase Inhibitor, Solid, 5 mg
Description
- Molecular Weight: 338.5 Da
- Purity: >98%
- Form: Solid
- Description: Tetracyclic diterpene antibiotic that acts as a reversible inhibitor of eukaryotic nuclear DNA replication by blocking DNA polymerase α, δ and ε.
- Biological Activity: Blocks the cell cycle at early S phase.
- Storage Temperature: Store at +4°C.
- Citation Count: 12
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | DNA Polymerase (Alpha), DNA Polymerase (Delta), DNA Polymerase (Epsilon) |
| Quantity | 5 mg |
| For Use With (Application) | DNA replication inhibition, Cell cycle blockade |
| Storage Requirements | Store at 4°C. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?