Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Aphidicolin, MP Biomedicals™

Catalog No. MP218387305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218387305 5 mg
MP218387301 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP218387305 Supplier MP Biomedicals, Inc Supplier No. 0218387305
Add to Cart
Add to Cart

By inhibiting the cellular replication of DNA polymerase II or the viral-induced DNA polymerases, Amphidocolin inhibits the growth of eukaryotic cells and certain animal viruses.

  • Aphidicolin is an antibiotic, a potent antiviral and antimitotic agent and DNA polymerase inhibitor
  • This compound is a featured product for Apoptosis research and in mitosis to research and track cellular activities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38966-21-1
Molecular Formula C20H34O4
Synonym aphidicolin, 38966-21-1, ICI 69653, (+)-Aphidicolin, (3 R,4 R,4 aR,6 AS,8 R,9 R,11 AS,11 bS)-4,9-bis(hydroxymethyl)-4,11 b-dimethyltetradecahydro-8,11 A-methanocyclohepta[a]naphthalene-3,9-diol

Specifications

Quantity 5 mg
Melting Point 226°C to 235°C
Color White
Purity >98
Molecular Weight (g/mol) 338.49
Chemical Name or Material (+)-Aphidicolin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.