Learn More
Selleck Chemical LLC Apigenin 50mg 520-36-5 NSC 83244, LY 080400
SDP
Supplier:  Selleck Chemical LLC S226250MG
Encompass_Preferred
Apigenin (NSC 83244, LY 080400) is a potent P450 inhibitor for CYP2C9 with Ki of 2 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2218
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.