Learn More
Apexbio Technology LLC Apixaban 503612-47-3 10mg
SDP
Supplier:  Apexbio Technology LLC A434110
Encompass_Preferred
Apixaban (CAS 503612-47-3) is a highly selective reversible inhibitor of coagulation Factor Xa with Ki values of 0 08 nM and 0 17 nM for human and rabbit enzymes respectively By targeting Factor Xa a central serine protease in the coagulation cascade apixaban effectively impedes thrombin generation and subsequent clot formation In vitro apixaban prolongs coagulation parameters in human plasma at submicromolar concentrations and demonstrates potent antithrombotic activity across multiple animal models With favorable pharmacokinetics including high oral bioavailability and low clearance in preclinical studies apixaban serves as a valuable tool for investigating coagulation mechanisms and evaluating anticoagulant strategies
Catalog No. 50-224-0731
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.