Learn More
Specifications
Specifications
| For Use With (Application) | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Product Type | APO-BRDU (TUNEL) Apoptosis Kit |
| Content And Storage | Negative Control Cells, Wash Buffer, Reaction Buffer, TdT Enzyme, Br-dUTP, Rinsing Buffer, Propidium Iodide/RNase DNA staining solution, Fluorescein-anti-BrdU antibody. Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
