Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ APO BrdU (TUNEL) Kit

Catalog No. 501121521
Encompass_Preferred
Change view
Click to view available options
Quantity:
60 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-1521 60 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-1521 Supplier Molecular Probes™ Supplier No. KHO1001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

APO BrdU (TUNEL) Kit

The ApoTarget™ APO-BRDU Kit is a two-color staining method for labeling DNA breaks and total cellular DNA to detect apoptotic cells by flow cytometry.

View a selection guide for all apoptosis assays for flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 60 tests
Format Tube
Product Type Apo-BRDU Tunel Kit
Content And Storage Contains pre-coated plate, standard, detector antibody, diluents, wash buffer, chromogen, stop solution and complete protocol
Flow Cytometer Laser Lines 488
Excitation/Emission FITC: 490/525
PI: 535/617
No. of Reactions 60
Color Green, Red
Product Line ApoTarget
Detection Method Fluorescence
For Use With (Application) Flow Cytometry
For Use With (Equipment) Flow Cytometer
Label or Dye FITC (Fluorescein), PI (Propidium Iodide)
Label Type Classic Dyes
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.