Learn More
R&D Systems™ APO-DIRECT (TUNEL) Apoptosis Kit
SDP
Catalog No. NBP231159 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
NBP231159 1 Kit
1 options
Catalog No. NBP231159 Supplier R&D Systems™ Supplier No. NBP231159
Customers require a valid business account associated to their profile to access pricing and ordering options.
A Novus product, part of R&D Systems. Applications: Flow Cytometry
Specifications
For Use With (Application) Flow Cytometry
Product Type APO-DIRECT (TUNEL) Apoptosis Kit
Content And Storage Positive Control Cells, Negative Control Cells, Wash Buffer, Reaction Buffer, TdT Enzyme, Rinsing Buffer, FITC dUTP, Propidium Iodide/RNase DNA staining solution. Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Line Novus
Quantity 1 Kit