Learn More
Sigma Aldrich Fine Chemicals Biosciences apo Transferrin human powd1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T4382
Encompass
Apo-transferrin human is a glycoprotein possessing homologous carboxyl and amino domain. Both the domain binds to one molecule of iron. Only 30% of transferrin is saturated with iron in the amino domain. Apo-transferrin in human is encoded by the gene TF which is localized on human chromosome 3q22.1.
Catalog No. 50-290-7989
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.