Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ApoA1 Monoclonal Antibody (6001)
GREENER_CHOICE

Catalog No. PIMA514667 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514667 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514667 Supplier Invitrogen™ Supplier No. MA514667
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

By sandwich ELISA, MA5-14667 can be used as a detection antibody with Product # 710263 as the coating antibody, to generate a matched pair. Using this matched pair, recombinant human Apo A-1, but not recombinant mouse Apo A-1, was detected. It is not recommended to use Product # 701239 for coating when MA5-14667 is used for detection. By sandwich ELISA, MA5-14667 can be used to detect Apo A-1 from serum samples. To increase sensitivity of sandwich ELISAs with MA5-14667, a biotinylated detection antibody followed by Streptavidin-HRP is recommended. By Western blot, MA5-14667 detects recombinant human Apo A-1, but not recombinant mouse Apo A-1. MA5-14667 may not be as successful for detecting endogenous Apo A-1 by Western blot. Product MA514667 is a smaller package size of MIA1401 (formerly sold as a Seradyn product).

Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ApoA1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 6001
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene APOA1
Gene Accession No. P02647
Gene Alias 1-Sep; 2-Sep; Alp-1; Apo a; APO A1; apo A-1; APO I; apo(a); APOA1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; apolipoprotein A-I; apolipoprotein A-I precursor; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; prepro-apolipoprotein-AI; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7
Gene Symbols APOA1
Host Species Mouse
Immunogen Purified human plasma Apo A-1.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 335
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.