Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOA1BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560502 Supplier Invitrogen™ Supplier No. PA560502
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQTIACRSGP TWWGPQRLNS GGRWDSEVMA STVVKYLSQE EAQAVDQELF NEYQFSVDQL MELAGLSCAT AIAKAYPPTS MSRS Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 75%.

The product of this gene interacts with apolipoprotein A-I (apoA-I), the major apolipoprotein of high-density lipoproteins (HDLs). It is secreted into some bodily fluids, and its synthesis and secretion are stimulated in vitro by incubating cells with apoA-I. The human genome contains related pseudogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOA1BP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAXE
Gene Accession No. Q8NCW5
Gene Alias AA087124; AIBP; AI-BP; AI-BP {ECO:0000255; Apoa1bp; Apoa1ip; apoA-I binding protein; apolipoprotein A-I binding protein; apolipoprotein A-I interacting protein; apolipoprotein A-I-binding protein; apolipoprotein A-I-binding protein {ECO:0000255; ESTM37; HAMAP-Rule:MF_03159}; MGC119143; MGC119144; MGC119145; NAD(P)H-hydrate epimerase; NAD(P)H-hydrate epimerase {ECO:0000255; NAD(P)HX epimerase; NAD(P)HX epimerase {ECO:0000255; NAXE; YjeF N-terminal domain-containing protein 1; yjeF_N1; YJEFN1
Gene Symbols NAXE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APOA1BP. Recombinant protein control fragment (Product #RP-100416).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128240
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.