Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOB48R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559864
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559864 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559864 Supplier Invitrogen™ Supplier No. PA559864
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGCGTEEGEA SVSENQELDG STGADAGPCP SLGEAYARET EDEEAEADRT SRRGWRLQAV AVGLPDREDA QTGSVAAGIM GGDVVPHISA AGAGEALEGV Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 36%.

Macrophage receptor that binds to the apolipoprotein B48 (APOB) of dietary triglyceride (TG)-rich lipoproteins (TRL) or to a like domain of APOB in hypertriglyceridemic very low density lipoprotein (HTG-VLDL). Binds and internalizes TRL when out of the context of the macrophage. May provide essential lipids to reticuloendothelial cells. Could also be involved in foam cell formation with elevated TRL and remnant lipoprotein (RLP). Mediates the rapid high-affinity uptake of chylomicrons (CM), HTG- VLDL, and trypsinized (tryp) VLDL devoid of APOE in vitro in macrophages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOB48R
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APOBR
Gene Accession No. Q0VD83
Gene Alias APOB100R; Apob48r; apoB-48R; Apobr; Apolipoprotein B receptor; apolipoprotein B100 receptor; apolipoprotein B-100 receptor; apolipoprotein B48 receptor; Apolipoprotein B-48 receptor; RGD1565169
Gene Symbols APOBR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APOB48R. Recombinant protein control fragment (Product #RP-98786).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55911
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.