Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOBEC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA511429
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA511429 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA511429 Supplier Invitrogen™ Supplier No. PA511429
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APOBEC1 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the cytidine deaminase enzyme family. The encoded protein forms a multiple-protein editing holoenzyme with APOBEC1 complementation factor and APOBEC1 stimulating protein . This holoenzyme is involved in the editing of C-to-U nucleotide bases in apolipoprotein B and neurofibromatosis-1 mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOBEC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene APOBEC1
Gene Accession No. P41238, P51908
Gene Alias Apobec1; APOBEC-1; apolipoprotein B editing complex 1; apolipoprotein B mRNA editing enzyme catalytic polypeptide 1; apolipoprotein B mRNA editing enzyme catalytic subunit 1; apolipoprotein B mRNA editing enzyme complex-1; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1; apolipoprotein B mRNA-editing enzyme 1; BEDP; C->U-editing enzyme APOBEC-1; Cdar1; HEPR; mRNA(cytosine(6666)) deaminase 1
Gene Symbols APOBEC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 7-36 amino acids from the N-terminal region of human Apobec1.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11810, 339
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.