Learn More
Apolipoprotein AI Mouse anti-Human, Mouse, Clone: 1C9G5, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody was generated against the C-terminal region of human ApoA1.
Specifications
Specifications
| Antigen | Apolipoprotein AI |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 1C9G5 |
| Concentration | 1.07 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | APOA1 |
| Gene Accession No. | P02647, Q00623 |
| Gene Alias | Apo AI, ApoA I, APOA1, apolipoprotein A I, Apolipoprotein A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.