Learn More
CPC Scientific H-Lys-Tyr-Tyr-Glu-Leu-Glu-Glu-Lys-Ile-Val-Ser-Leu-Ile-Lys-Asn-Leu-Leu-Val-Ala-Leu-Lys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific APOL001B
Encompass
SEQUENCE: H-Lys-Tyr-Tyr-Glu-Leu-Glu-Glu-Lys-Ile-Val-Ser-Leu-Ile-Lys-Asn-Leu-Leu-Val-Ala-Leu-Lys-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: KYYELEEKIVSLIKNLLVALKMOLECULAR FORMULA: C119H200N26O32MOLECULAR WEIGHT: 2507STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Alzheimer's DiseaseREFERENCES:SKU(s): APOL-001A, APOL-001B
Catalog No. 50-210-8785
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.