Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein E/ApoE Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB001919
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001919 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001919 Supplier R&D Systems Supplier No. NBP131123FR

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Apolipoprotein E/ApoE Polyclonal specifically detects Apolipoprotein E/ApoE in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Apolipoprotein E/ApoE
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571
Gene Symbols APOE
Host Species Rabbit
Immunogen Apolipoprotein E/ApoE Antibody made to carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Apolipoprotein E. The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Apoptosis, Cancer, Cholesterol Metabolism, Cognition and Behavior, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 348
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.