Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein H Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4D9A4, Proteintech
SDP

Catalog No. 501736511 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6511 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6511 Supplier Proteintech Group Inc Supplier No. 660741IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.

Specifications

Antigen Apolipoprotein H
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4D9A4
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene APOH
Gene Accession No. P02749, P26644, Q01339
Gene Alias Anticardiolipin cofactor, APC inhibitor, Apo H, APOH, Apolipoprotein H, B2G1, B2GPI, Beta 2 glycoprotein 1, Beta 2 glycoprotein I, Beta(2)GPI, BG
Gene Symbols Apoh
Host Species Mouse
Immunogen Apolipoprotein H Fusion Protein Ag17915
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100522100, 11818, 287774, 350
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.