Learn More
ADELPHI MANUFACTURING Apollo II High Intensity Liquid Inspection Unit
Supplier:  ADELPHI MANUFACTURING 6031521
Encompass_Preferred
For coloured glass containers, translucent plastic containers, and turbid or coloured preparations.In front of the black panel, light coloured particulates will be visible, and in front of the white panel, dark coloured particulates will be visible.Compliant with US Pharmacopoeia USP chapter 41 (1790), section 6.1Height and angle adjustable.Intuitive assembly; no tools required.Light level between 8,000 and 10,000 Lux.Bulbs have a service life of 50,000 hours.Folds for storage, with the inspection surfaces together to prevent damage.Optional IQ/OQ documentation.
Catalog No. NC3825766
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.