Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Apoptosis Detection Antibody Pack
SDP

Catalog No. NBP225080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
3 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225080 3 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225080 Supplier Novus Biologicals™ Supplier No. NBP225080
Add to Cart
Add to Cart

Includes

NB100-56527 (Caspase-8 Antibody)-0.025mg, NB100-56599 (PARP Antibody)-0.025mg, NB100-56708 (Caspase-3 Antibody)-0.025mg

For Research applications

Apoptosis Detection specifically detects Apoptosis Detection in Human, Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen Apoptosis Detection
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot
Conjugate Unconjugated
Dilution Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready
Host Species Mouse
Immunogen The following antibodies are provided in Apoptosis Detection Antibody Bundle: 1. Caspase-3 (Pro and Active/Cleaved): 0.025mg: Localized to the large domain subunit: WB, IHC (paraffin), IHC (frozen): H, M, R: Western blot: pro and active forms seen 2. Caspase-8 (Pro and Active/Cleaved): 0.025mg: 370 PVETDSEEQP 379: WB: H, Ch, Rh: Western blot: pro and active forms seen 3. PARP (cleavage-specific): 0.025mg: A peptide containing amino acids near the 214/215-cleavage site: WB, Flow: Human: Western blot: specifically detects the cleaved form of PARP
Quantity 3 Vials
Test Specificity The Apoptosis Detection Antbody Bundle contains monoclonal antibodies to three key apoptosis markers: caspase-3, caspase-8, and cleaved PARP. The caspase-3 and -8 antibodies both recognize pro and cleaved forms. The PARP antibody specifically recognizes the 89 kDa PARP fragment and does not recognized the full-length protein.
Includes NB100-56527 (Caspase-8 Antibody)-0.025mg, NB100-56599 (PARP Antibody)-0.025mg, NB100-56708 (Caspase-3 Antibody)-0.025mg
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.