Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ ApoScreen™ Annexin V-APC
SDP

Catalog No. OB1004011 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1004011 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB1004011 Supplier SouthernBiotech™ Supplier No. 1004011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SouthernBiotech ApoScreen™ are valuable tools for your apoptosis research.

ApoScreen® Annexin V (SB Cat. No. 10040) is a recombinant protein that is expressed in E. coli. A number of changes in cell structure occur during apoptosis. One major change is the loss of membrane phospholipid asymmetry, with translocation of phosphatidylserine (PS) from the inner leaflet of the phospholipid bilayer to the external surface. Annexin V, a member of the annexin family of calcium-dependent phospholipid binding proteins, has a high affinity for phosphatidylserine (PS)-containing phospholipid bilayers. Conjugated Annexin V can be used to monitor changes in cell membrane phospholipid asymmetry, thereby providing a convenient tool for detection of apoptotic cells.

Specifications

Quantity 100 Tests
Storage Buffer Store at 2-8°C; Protect from light; Do not freeze
Content And Storage Supplied in PBS/NaN3 and a stabilizing agent
For Use With (Application) Flow Cytometry
Conjugate APC (Allophycocyanin)

For Research Use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.