Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ ApoScreen™ Annexin V Binding Buffer 10X
SDP

Catalog No. OB1004501 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1004501 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB1004501 Supplier SouthernBiotech™ Supplier No. 1004501
Add to Cart
Add to Cart

SouthernBiotech ApoScreen™ are valuable tools for your apoptosis research.

ApoScreen® Annexin V Binding Buffer 10X has been formulated for flow cytometric labeling of apoptotic cells with ApoScreen® Annexin V reagents.

Specifications

Quantity 50 mL
Storage Buffer Supplied as a saline solution with a stabilizing agent and 0.1% NaN3 as a preservative
Content And Storage 2°C to 8°C
For Use With (Application) Flow Cytometry

For Research Use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.