Learn More
Andwin Scientific APPLICATORS 1000/PK. NO GLUE
SDP
Supplier:  Andwin Scientific 10806005
Encompass_Preferred
The Cotton-Tipped Wooden Applicators are efficient product for use in bacteriology labs for inoculating broth and plated media, and obtaining culture specimens. The applicators can also be utilized for sanitizing delicate laboratory equipments or instruments. The wooden handle measures 152.4 mm (6.00") x 2.5 mm (0.100") while the head measures 15.9 mm (0.625") x 4.7 mm (0.188"). This item is sold in a pack of 1000.
Catalog No. NC9538864
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.