Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ 8-Cap Strip, clear

Catalog No. p-4926358
Encompass_Preferred
Change view
Click to view available options
Quantity:
1500 Strips (5 pk./300)
300 Strips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8010535 300 Strips
N8011535 1500 Strips (5 pk./300)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N8010535 Supplier Applied Biosystems™ Supplier No. N8010535
Only null left
Add to Cart
Add to Cart

Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates

MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 8-cap strips available in clear or an assorted set of red, orange, blue, and green.

  • Will not interfere with sample reading
  • Provides tight seal to minimize evaporation
  • Easy to apply and remove
Minimizes Evaporation
  • Use MicroAmp Caps Strips to seal tubes and reduce the possibility of contamination in sample wells and help ensure reliable results
  • Easy to Use
    • Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
    • When ready, just peel away caps using tabs at end
    Colored for Easy Organization
    • Available in different colors to help track tubes during assay preparation
    • For real-time PCR applications,use MicroAmp Optical 8-Cap Strips (Mfr. No. 4323032) with flat-top profile

    PCR, PCR & Real-Time PCR

    TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance Certified DNA/Rnase Free
    Color Natural
    Quantity 300 Strips
    For Use With (Equipment) Thermal Cyclers
    Product Line MicroAmp™ 
    Material Polypropylene
    Content And Storage Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates. Natural (clear) color. Store at room temperature.
    Shipping Condition Room Temperature
    Closure Type Domed Cap
    No. of Wells 96
    Format 300 Strip Caps
    Product Type 8-Strip Cap
    Show More Show Less
    WARNING: Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.