Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Fast Reaction Tube with Cap, 0.1 mL

Catalog No. 4358297
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-582-97 1000 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-582-97 Supplier Applied Biosystems™ Supplier No. 4358297
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Single reaction tube, with flat cap

Requires

MicroAmp 96-Well Tray for Veriflex™ Blocks (Mfr. No. 4379983)

Reduce PCR reaction time to as little as 25 minutes

The complete Applied Biosystems fast system, comprising premixed Fast PCR Master Mixes, specially-designed 0.1mL microplates and tubes, and the Veriti 96-Well Fast Thermal Cycler, considerably reduces your PCR reaction times. Now, you can amplify fragments under 500 bp from genomic backgrounds in about 25 minutes and, in some cases, as little as 10 minutes. And you can amplify large fragments faster too—up to 2kb in as little as 40-50 minutes. Or for sequencing applications, the AmpliTaq Gold™ Fast PCR Master Mix, UP reduces your cycling time to 50 minutes and produces amplicons optimal for high quality sequencing results.

  • Fits snugly in Veriti™ 96-Well Fast Thermal Cycler to maximize thermal conductivity and provide precise control of cycling temperatures
  • Enhances thermal conductivity, enabling precise control of cycling temperatures
  • Prevents wasted wells when you only have few samples to run
  • Ensures reproducible, specific, and sensitive results
  • Reduces PCR reaction time from 2 hours to as little as 25 minutes when used with the Veriti 96-Well Fast PCR System
  • Stop Wasting Wells
  • If you have only few samples to run, use individual MicroAmp Fast Reaction Tubes with caps
  • Design of Fast Reaction Tubes maximizes thermal conductivity when used with Veriti 96-Well Fast Thermal Cycler
  • This enhanced thermal conductivity facilitates precise control of cycling temperatures
  • Part of Applied Biosystems Fast PCR System
    • Comprising premixed GeneAmp™ Fast PCR Master Mix, specially-designed microplates and tubes, and the Veriti 96-Well Fast Thermal Cycler, considerably reduces your PCR reaction times
    • Amplify in as little as 25 minutes

    Fast PCR, Fast Real-Time PCR, PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/Rnase Free
Color Natural
Volume (Metric) Working 0.1 mL
Volume (Metric) 0.15 mL
Quantity 1000 Tubes
For Use With (Equipment) Thermal Cyclers
Product Line MicroAmp™ 
Material Polypropylene
Content And Storage Single reaction tube, with flat cap, for 9800 Fast PCR System. Use with Fast 96-Well Retainer Tray (4358305).
Store at RT
Reaction Speed Fast
Shipping Condition Room Temperature
Closure Type Flat Cap
No. of Wells 96
Format 1,000 Tubes
Product Type Reaction Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.