Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Optical 8-Cap Strips

Catalog No. 4323032
Encompass_Preferred
Change view
Click to view available options
Quantity:
300 Strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-230-32 300 Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-230-32 Supplier Applied Biosystems™ Supplier No. 4323032
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Tight seal minimizes evaporation and contamination

Designed with flat-top profile to fit on MicroAmp tube strips and 96-well plates. Suitable for real-time PCR.

  • Will not interfere with sample reading
  • Provides tight seal to minimize evaporation
  • Easy to apply and remove
  • Minimize Evaporation
  • Use MicroAmp Caps Strips to seal the tubes and reduce the possibility of contamination among sample wells and help ensure reliable results
  • Easy to Use
  • Use MicroAmp Cap Installing Tool (sold separatley) to push caps down and form tight seal. When ready, just peel away caps using tabs at end
  • Optical caps have been redesigned with improved sealing geometry while still featuring excellent optical clarity and low fluorescent background

For end-point PCR applications, use MicroAmp Cap Strips with domed-top profile.

PCR and Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/Rnase Free
Color Optical
Volume (Metric) 0.2 mL
Quantity 300 Strips
For Use With (Equipment) Thermal Cyclers or Real-Time PCR Systems
Product Line MicroAmp™ 
Material Polypropylene
Content And Storage Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates.
Reaction Speed Fast
Shipping Condition Room Temperature
Closure Type Optically Clear Flat Caps
No. of Wells 48/96
Format 300 Strip Tubes
Product Type Optical 8-Tube Strip
Show More Show Less
Can I run strip tubes on the ViiA 7 instrument?

Yes. Make sure to use the appropriate matching parts. For the 96-well standard block, use the standard 96-well plate adapter (Cat. No. 4459845), the retainer tray for (0.2 mL) tubes, and Optical 8-Tube Strips and Caps (Cat. No. 4316567 and 4323032). For the 96-well Fast block, use the Fast 96-well plate adapter (Cat. No. 4462078), the retainer tray (Cat. No. 4379983), and Fast (0.1 mL) Optical 8-tube Caps and Strips (Cat. Nos. 4323032 and 4358293).

Which of your plastic consumables are compatible with the Applied Biosystems 7900HT Fast Real-Time PCR System?

There are many of our reagents and plastic consumables that can be used on the Applied Biosystems 7900HT Fast Real-Time PCR System. Here are a few examples to help you get started:
Cat. No. 4313663, MicroAmp Optical Adhesive Film Kit
Cat. No. 4311971, 4360954, MicroAmp Optical Adhesive Film
Cat. No. 4323032, MicroAmp Optical 8-Cap Strip
Cat. No. 4309849; 4326270, MicroAmp Optical 384-Well Reaction Plate with Barcode
Cat. No. 4306737; 4326659; 4314320, MicroAmp Optical 96-Well Reaction Plate with Barcode
Cat. No. 4346906, MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Cat. No. 4312063, Splash Free 96-Well Base

For more comprehensive lists of compatible plastics, enter "Real-Time PCR Plastics" in the main Search field on our website and follow the links.

What is the difference between the MicroAmp Optical Adhesive Covers and the MicroAmp Optical Caps? Can they be used interchangeably?

The MicroAmp Optical Adhesive Covers (P/N 4311971) have a higher optical efficiency than the MicroAmp Optical Caps, (8 caps/strip) (P/N 4323032). They can be used instead of the MicroAmp Optical Caps only when using the MicroAmp 96-Well Reaction Plate with Barcode (P/N 4306737, 4309849) or the MicroAmp Optical 96-Well Reaction Plate (P/N N801-0560). The Optical Adhesive Covers cannot be used with the MicroAmp Optical Tubes.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.