Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Adhesive Films

Catalog No. 4314320
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-143-20 100 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-143-20 Supplier Applied Biosystems™ Supplier No. 4314320
  • $1,326.00
    $1,259.65 / Each of 1

    Save $66.35

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

5 packages of 20 plates each (equivalent to 5 x Mfr. No. 4306737), 1 package of 100 Optical Adhesive Films (equivalent to Mfr. No. 4311971)

Optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification

Constructed from a single rigid piece of polypropylene in a 96-well format, this plate is compatible with Applied Biosystems 96-Well Real-Time PCR systems and thermal cyclers.

  • Frosted to minimize interfering fluorescence from wells of cycling block
  • Screened to eliminate autofluorescent plates
  • Unique design provides a barrier to ambient air to help insure well-to-well temperature uniformity
  • Each reaction plate includes a unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
  • The included MicroAmp Optical Adhesive Films reduce the chance of well-to-well contamination and sample evaporation when applied to the microplates
  • Pressure-sensitive adhesive provides a tight seal over every well
  • Will not interfere with sample reading
  • Confirm correct alignment to plate, then press to seal
  • Easy to apply; does not stick to glove
  • Capcity: 96 x 0.2mL tubes
Oil-Free PCR Amplification
  • MicroAmp disposables are optimized to provide unmatched temperature accuracy and uniformity for fast, oil-free PCR amplification

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Ambient

TRUSTED_SUSTAINABILITY

Specifications

Color Optical
Certifications/Compliance Certified DNA/RNase Free
Material Polypropylene
No. of Wells 96
Quantity 100 plates
Content And Storage Contains:
5 packages of 20 plates each (equivalent to 5 × catalog # 4306737)
1 package of 100 Optical Adhesive Films (equivalent to catalog # 4311971)

Store at room temperature. With proper storage, shelf-life is 2 years.
Format 96-well plate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Product Type Plate/Adhesive Film/Barcode
Product Line MicroAmp
Adhesive Pressure Sensitive
Plate Compatibility 96-well Plate
Show More Show Less
Can I place a standard MicroAmp 96-Well Optical Reaction Plate onto a 7500 Fast Real-Time PCR System?

No. The plate does not fit and will result in the crushing of the wells and possible contamination of the block due to leaking of the well contents.

Which of your plastic consumables are compatible with the Applied Biosystems 7900HT Fast Real-Time PCR System?

There are many of our reagents and plastic consumables that can be used on the Applied Biosystems 7900HT Fast Real-Time PCR System. Here are a few examples to help you get started:
Cat. No. 4313663, MicroAmp Optical Adhesive Film Kit
Cat. No. 4311971, 4360954, MicroAmp Optical Adhesive Film
Cat. No. 4323032, MicroAmp Optical 8-Cap Strip
Cat. No. 4309849; 4326270, MicroAmp Optical 384-Well Reaction Plate with Barcode
Cat. No. 4306737; 4326659; 4314320, MicroAmp Optical 96-Well Reaction Plate with Barcode
Cat. No. 4346906, MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Cat. No. 4312063, Splash Free 96-Well Base

For more comprehensive lists of compatible plastics, enter "Real-Time PCR Plastics" in the main Search field on our website and follow the links.

Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?

No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.