Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Optical Film Compression Pad

Catalog No. 4312639
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-126-39 5 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-126-39 Supplier Applied Biosystems™ Supplier No. 4312639
  • $101.00
    $96.65 / Each of 1

    Save $4.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

5 Optical Cover Compression Pads

For use with adhesive films

Designed to support a proper thermal seal between the thermal cycler and reaction plates when using adhesive films.

See user's manual or package insert for limited label license, and trademark information.

PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Optical
Quantity 5 Ea.
For Use With (Application) PCR
Content And Storage 5 Optical Cover Compression Pads
For Use With (Equipment) 2720 Thermal Cycler, 3100 Genetic Analyzer, 3700 Genetic Analyzer, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 System, 7900HT System, GeneAmp 9700, Veriti Thermal Cycler, 2720 Thermal Cycler
Shipping Condition Room Temperature
Product Type Compression Pad
Product Line MicroAmp
Plate Compatibility 96-well Plate
Can I use an MicroAmp Optical Cover Compression Pad on an Optical Fast 96-Well plate?

No. we do not recommend placing compression pads on the Fast 96-well plate or the StepOnePlus, ViiA 7, 7300, 7500, 7900HT, or 7500 Fast Real-Time PCR Systems.

Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?

No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.

How many times can a MicroAmp Optical Film Compression Pad (Cat. No. 4312639) be used?

With proper usage, the compression pad should maintain performance for a minimum of 20 PCR runs. It is important to examine the pad before each run, especially after more than 20 uses, as deteriorated compression pads may lead to evaporation and failed reactions.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.