Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MUSTANG PURPLE™ Dye Spectral Calibration Plate, 96-well

Catalog No. 4461599
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-615-99 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-615-99 Supplier Applied Biosystems™ Supplier No. 4461599
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for MUSTANG PURPLE™ spectral calibration of your 7500, ViiA™ 7, QuantStudio™ 6, QuantStudio™ 7, or QuantStudio™ 12K Flex RT-PCR system with a 96-well block.

The dye formulations will help improve multiplexing results by more accurately representing fluorescent spectra that are part of your real-time PCR experiments.

See instrument maintenance guide for full instructions. The dye formulation will help improve multiplexing results by more accurately representing fluorescent spectra that are part of real-time PCR experiments.

Order Info

Dry ice shipping; store in freezer (-5° to -30°C)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in Freezer (-5°C to -30°C).
Calibrated Dye Mustang Purple™
For Use With (Equipment) 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System, QuantStudio™ 7, QuantStudio™ 6, QuantStudio™ 12K
Product Type Dye Spectral Calibration Plate
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 plate
Calibration Type Spectral Calibration
Do you offer MUSTANG PURPLE Dye Spectral Calibration Plate, 96-well (Cat. No. 4461599) as a stand-alone passive reference dye for purchase?

No, we do not offer Mustang Purple as a stand-alone product.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.