Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Platinum™ Multiplex PCR Master Mix

Catalog No. 4464270
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
2000 Reactions
250 Reactions
50 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
44-642-70 2000 Reactions
44-642-68 50 Reactions
44-642-69 250 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 44-642-70 Supplier Applied Biosystems™ Supplier No. 4464270
Only null left
Add to Cart
Edge
Add to Cart
Request a Quote

Includes

Platinum Multiplex PCR Master Mix, 2X; GC Enhancer

The Platinum Multiplex PCR Master Mix is designed specifically for endpoint multiplex PCR.

Platinum Multiplex PCR Master Mix is a high-specificity, high-throughput solution designed specifically for endpoint multiplex PCR and supports easy multiplexing with minimal optimization.

The performance of the Platinum Multiplex PCR Master Mix over a wide range of amplicon sizes permits the amplification of templates from 50 bp to 2.5 kb, greatly enhancing workflow flexibility. Coupled with its 20-plex capability and absence of primer dimers, it not only provides a high-throughput solution but also boasts high specificity through fewer non-specific primer binding events, resulting in less reaction and primer waste.

Features

  • Amplifies up to 20 amplicons in a single reaction
  • Amplifies products from 50 bp to 2.5 kb
  • Performs multiplex PCR reactions with minimal optimization

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • Platinum Multiplex PCR Master Mix, 2X (5 x 10 mL)
• GC Enhancer (1 x 10 mL)

Store at –15°C to –25°C prior to opening.
Master mix may be stored at –15°C to –25°C or at 4°C (for up to 30 days). The GC Enhancer must be kept at –15°C to –25°C.
GC-Rich PCR Performance Low
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Standard
Multiplex Capability Up to 20-plex
Quantity 2000 Reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, Multiplex PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 2000 Reactions
Reaction Format SuperMix or Master Mix
Size (Final Product) 2.5 kb or less
Volume 10 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.