Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ StepOnePlus™ Real-Time PCR System Spectral Calibration Kit (96 well)

Catalog No. 4371435
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-714-35 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-714-35 Supplier Applied Biosystems™ Supplier No. 4371435
  • $1,064.00
    $1,031.65 / Each of 1

    Save $32.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Includes one background plate and two 96-well fast reaction plates containing spectra dyes (plates I & II). Plate I contains FAM, VIC, NED, and ROX; Plate II contains SYBR Green I, JOE, and TAMRA.

Easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™/SYBR™ Green I, VIC™/JOE™, and NED™/TAMRA™/ROX™ dyes on the StepOnePlus™ RT-PCR System.

The Applied Biosystems™ StepOnePlus™ Real-Time PCR System Spectral Calibration Kit contains one background plate and two preloaded, 96-well fast optical reaction plates containing seven dye standards.

This easy-to-use kit enables users to calibrate the StepOnePlus Real-Time PCR System quickly and easily. Insert the reaction plates during system installation, and the system automatically collects the dye standards' spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.

  • Provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture
  • Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions

Order Info

Dry ice shipping; store at -15° to -25°C; protect from light

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Includes one background plate and two 96-well fast reaction plates containing spectra dyes (plates I and II). Plate I contains FAM, VIC, NED, and ROX; Plate II contains SYBR Green I, JOE, and TAMRA.

Store at -15°C to -25°C. Protect from light.
Calibrated Dye FAM, JOE, NED, ROX, TAMRA™, VIC, SYBR™ Green
For Use With (Equipment) Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode
Detection Method Fluorescence
Product Line StepOnePlus
Product Type Spectral Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 Kit
Calibration Type Spectral Calibration
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.