Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ System 9700 Networking Software Expansion Kit

Catalog No. 4332266
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-322-66 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-322-66 Supplier Applied Biosystems™ Supplier No. 4332266
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Software update for 4332267, 9700 network software expansion kit.

Requires

233 MHz processor or better, 60MB free hard disk space, 128MB RAM, Microsoft Windows 95/98, NT, 2000

Expands existing System 9700 Networking Software

Used With: 0.5mL GeneAmp™ PCR System 9700, Aluminum 96-Well GeneAmp PCR System 9700, Auto-Lid Dual 384-Well GeneAmp PCR System 9700, Dual 384-Well GeneAmp PCR System 9700, Gold-plated 96-Well GeneAmp PCR System 9700, Silver 96-Well GeneAmp PCR System 9700, System 9700 Networking Software

Gene Expression Analysis & Genotyping, Genotyping & Genomic Profiling, PCR, PCR & Real-Time PCR

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Software update for 4332267, 9700 network software expansion kit.
For Use With (Equipment) GeneAmp 9700
Software Type Software
Product Type Software Expansion Kit
For Use With (Application) PCR
Shipping Condition Room Temperature
Operating System Windows 2000, Windows 98, Windows 95, Windows NT
Quantity 1 kit
Memory 128 MB RAM

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.