Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ RNase P Instrument Verification Plate, Fast 48-well

Catalog No. 4371439
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 48-well plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-714-39 1 x 48-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-714-39 Supplier Applied Biosystems™ Supplier No. 4371439
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Contains 48-well Fast Reaction Plate preloaded and sealed with TaqMan™ RNase P probes and primers.

Used to verify the performance of the Applied Biosystems™ StepOne™ Real-Time PCR System.

The TaqMan™ RNase P Fast 48-well Instrument Verification Plate is a 48-well PCR plate preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye–labeled probe) and template. The TaqMan™ RNase P Fast 48-well Instrument Verification Plate satisfies the same requirements as the Applied Biosystems™ StepOne™ Real-Time PCR Systems installation. To pass installation, the instrument must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, Applied Biosystems™ recommends that the performance of your instrument be verified every six months.

The TaqMan RNase P Fast 48-Well Instrument Verification Plate satisfies the same requirements as the Applied Biosystems StepOne Real-Time PCR Systems installation. To pass installation, the instrument must demonstrate the ability to distinguish between 5000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, the performance of the instrument should be verified every six months.

Order Info

Dry ice shipping; store at -15° to 25°C; product is light sensitive

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 48-Well Fast Reaction Plate preloaded and sealed with TaqMan™ RNase P probes and primers. Store at -15 to 25°C. This product is light sensitive.
Calibrated Dye FAM
For Use With (Equipment) StepOne™, Fast Mode
Detection Method Fluorescence
Product Line TaqMan
Product Type Instrument Verification Plate
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 x 48-well plate
Calibration Type RNase P Calibration, Instrument Verification Plate
Can I use my 7900HT System RNase P instrument verification plates with both the 7900HT System and the QuantStudio 6 and 7 Flex Real-Time PCR Systems?

Only the TaqMan RNase P Instrument Verification Plate, Fast 96-well (for 0.1 mL block) (Cat. No. 4351979) is compatible with both the 7900HT System and the QuantStudio 6 and 7 Flex Real-Time PCR Systems. The following RNase P instrument verification plates can be used on the 7900HT System, but not on the QuantStudio 6 and 7 Flex Real-Time PCR Systems:
• TaqMan RNase P Instrument Verification Plate, 96-well (Cat. No. 4310982) (for 96-well standard block)
• TaqMan RNase P Instrument Verification Plate for 7900HT System, 384-well (Cat. No. 4323306) (for 384-well block)
• 7900HT TaqMan Low Density Array Instrument Verification RNase P Kit (Cat. No. 4351468) (for TaqMan Array Card block)

The following RNase P instrument verification plates can be used on the QuantStudio 6 and 7 Flex Real-Time PCR Systems, but not on the 7900HT System:
• TaqMan RNase P Instrument Verification Plate for ViiA7 System, 96-well (Cat. No. 4432382) (for 96-well standard block)
• TaqMan RNase P Instrument Verification Plate for ViiA7 System, 384-well (Cat. No. 4455280) (for 384-well block)
• ViiA7 Array Card RNase P Verification Kit (Cat. No. 4432265) (for TaqMan Array Card block)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.