Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ RNase P Instrument Verification Plate for ViiA™ 7 System, 384-well

Catalog No. 4455280
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 384-well plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-552-80 1 x 384-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-552-80 Supplier Applied Biosystems™ Supplier No. 4455280
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Used to perform verification of your ViiA™7 System with 384 Well Block.

Used to perform verification of your ViiA™7 System with 384 Well Block.

Please see your instrument maintenance guide for full instructions on use.

Order Info

Dry ice shipping; store in freezer (-5° to -30°C)

TRUSTED_SUSTAINABILITY

Specifications

Format 384-well Plate
Content And Storage Store Calibration Plates at -15°C to -25°C
Calibrated Dye FAM
For Use With (Equipment) ViiA™ 7 System, QuantStudio™ 5, QuantStudio™ 6 Pro, QuantStudio™ 7 Pro, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio™ 12k Flex
Product Line TaqMan, ViiA
Product Type Rnase P Instrument Verification Plate
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 x 384-well plate
Calibration Type RNase P Calibration, Instrument Verification Plate
When do I need to run the RNaseP calibration?

The RNaseP verification plate contains template, master mix, and a TaqMan assay for RNaseP. It is used to verify that the instrument is performing to specifications. If you have reason to suspect there is something wrong with the instrument, or if you want to rule out a chemistry issue, the RNaseP plate is a good way to test the system. The RNaseP verification plate is a single-use plate.

When do I need to run the RNaseP calibration for the QuantStudio 12K Flex Real-Time PCR System?

The RNaseP verification plate contains template, master mix, and a TaqMan assay for RNaseP. It is used to verify that the instrument is performing to specifications. If you have reason to suspect there is something wrong with the instrument, or if you want to rule out a chemistry issue, the RNaseP plate is a good way to test the system. The RNaseP verification plate is a single-use plate.

I cannot open the QuantStudio 3/5 Design & Analysis 2 (DA2) software on my Mac. What are the software requirements for Mac?

To open the software download on your Mac, you will need to adjust your security settings using the following instructions from Apple support:

Safely open apps on your mac

Please note, the software should run on Mac OS 10.10 or later.

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

Can I import a standard curve for my real-time PCR data into the QuantStudio Real-Time PCR Software?

If you are analyzing results from the ViiA 7 Real-Time PCR System, QuantStudio 6 Flex Real-Time PCR System, or QuantStudio 7 Flex Real-Time PCR System, it is possible to import a standard curve into the QuantStudio Real-Time PCR Software following the instructions below:
1. Open the data file in the QuantStudio Real-Time PCR Software.
2. Go to Analysis Settings.
3. Click on the "Standard Curve Settings" tab.
4. Select the option to "Use a standard curve imported from another experiment" and click on "Import".
5. Browse to and highlight the proper data file and click "Select".
6. Click Apply Analysis Settings.

Can I use my 7900HT System RNase P instrument verification plates with both the 7900HT System and the QuantStudio 6 and 7 Flex Real-Time PCR Systems?

Only the TaqMan RNase P Instrument Verification Plate, Fast 96-well (for 0.1 mL block) (Cat. No. 4351979) is compatible with both the 7900HT System and the QuantStudio 6 and 7 Flex Real-Time PCR Systems. The following RNase P instrument verification plates can be used on the 7900HT System, but not on the QuantStudio 6 and 7 Flex Real-Time PCR Systems:
• TaqMan RNase P Instrument Verification Plate, 96-well (Cat. No. 4310982) (for 96-well standard block)
• TaqMan RNase P Instrument Verification Plate for 7900HT System, 384-well (Cat. No. 4323306) (for 384-well block)
• 7900HT TaqMan Low Density Array Instrument Verification RNase P Kit (Cat. No. 4351468) (for TaqMan Array Card block)

The following RNase P instrument verification plates can be used on the QuantStudio 6 and 7 Flex Real-Time PCR Systems, but not on the 7900HT System:
• TaqMan RNase P Instrument Verification Plate for ViiA7 System, 96-well (Cat. No. 4432382) (for 96-well standard block)
• TaqMan RNase P Instrument Verification Plate for ViiA7 System, 384-well (Cat. No. 4455280) (for 384-well block)
• ViiA7 Array Card RNase P Verification Kit (Cat. No. 4432265) (for TaqMan Array Card block)

How can I get a summary of the calibrations performed on the QuantStudio 6 and 7 Flex Real-Time PCR Systems?

You can get a summary of the calibrations from the QuantStudio Software by going to Instrument > Instrument Console > Manage Instrument > Information > Print Calibration Log.

Do I need to back up my calibration files before I upgrade the software for the ViiA7 Real-Time PCR System?

No, you do not need to back up the calibration files as all of the calibration files are stored on the ViiA7 Real-Time PCR instrument. Once the computer software is properly installed and the computer is connected to the instrument, the calibration information will automatically update in the QuantStudio Software.

Can I still start a run on the ViiA7 Real-Time PCR System if the "Calibration is expired" notification is displayed?

Yes, you can still start a run on the ViiA7 Real-Time PCR System if the expired calibration notice displays. This is just a reminder and you can close it to proceed with the run. Additionally, you can change when you want the notifications to show up and how frequently you want to be notified to calibrate the instrument. To do this, go to the toolbar in the QuantStudio Software and select Instrument > Instrument Console. Select your instrument icon and click on Manager Instrument > Maintenance > Calibration Reminders. From the Calibration reminders Settings page you can update when the calibrations expire, turn expiration reminders on/off, and set when the reminders show up.

What is the purpose of the RNase P Detection Reagents Kit (Cat. No. 4316831), and what is provided in the kit?

RNase P is a single-copy gene encoding the RNA moiety for the RNase P enzyme, and it is an excellent gene to use as a reference/control for Copy Number Variation (CNV) assays. The TaqMan RNase P Detection Reagents Kit contains a 20X mix of primers and probe (FAM dye-labeled) that will detect and quantitate genomic copies of the human RNase P gene. The primers and probe are designed according to TaqMan chemistry guidelines for quantitation, and utilize the universal thermal cycling parameters. This kit is used to perform a 5' nuclease assay with TaqMan Universal PCR Master Mix (Cat. No. 4304437) or the TaqMan Core PCR Reagents Kit (Cat. No. N808-0228) with genomic, plasmid or complementary DNA (cDNA) templates.

The complete components of the TaqMan RNase P Detection Reagents Kit, Cat. No. 4316831, are: 20X RNase P Primer-Probe (FAM/TAMRA probe) Mix, 1 tube Human Genomic Control DNA, 10 ng/ul (100 ul), and the Product Insert with an overview and procedure for the use of the kit.


WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.